Boron tribromide 1 M in dichloromethane, AcroSeal®

Supplier: Thermo Scientific Chemicals
Danger

427101000
TS427101000EA 166 USD
TS427101000
Boron tribromide 1 M in dichloromethane, AcroSeal®
Boron tribromide
Formula: BBr₃
MW: 250.52 g/mol
Density: 1.467 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011312
CAS Number: 10294-33-4
EINECS: 233-657-9
UN: 3390
ADR: 6.1,

Order Now

Specification Test Results

Appearance Clear yellow to brown Solution (May develop some turbidity or precipitate. May darken on storage)
Strength 0.95 to 1.10 M
Titration Complexometric ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR