Hydrogen chloride 1.25 M in ethanol, AcroSeal®

Supplier: Thermo Scientific Chemicals
Danger

427011000 427018000
TS427011000EA 59 USD
TS427011000 TS427018000
Hydrogen chloride 1.25 M in ethanol, AcroSeal®
Hydrogen chloride
Formula: HCl
MW: 36.46
Flash Pt: 12 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 2924
ADR: 3 II

Order Now

Specification Test Results

Appearance Clear colorless Solution
Water ≤0.5 %
Strength 1.10 to 1.40 M
NMR ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR