Chloroform-[D₁] (99.8% D) + 0.03% (v/v) TMS, AcroSeal™ for NMR spectroscopy

Supplier: Thermo Scientific Chemicals
Danger

42696-1000
TS42696-1000EA 88 USD
TS42696-1000
Chloroform-[D₁] (99.8% D) + 0.03% (v/v) TMS, AcroSeal™ for NMR spectroscopy
Chloroform-[D1]
Formula: CDCl₃
Boiling Pt: 60.8 °C (1013 hPa)
Melting Pt: –64 °C
Density: 1.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000827
CAS Number: 865-49-6
EINECS: 212-742-4
UN: 1888
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.02 %
D-Enrichment ≥99.75 %
TMS 0.028 to 0.032 v/v%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR