Deuterium oxide 99.8 atom % D, AcroSeal™ for NMR spectroscopy

Supplier: Thermo Scientific Chemicals

42693-1000
TS42693-1000EA 190 USD
TS42693-1000
Deuterium oxide 99.8 atom % D, AcroSeal™ for NMR spectroscopy
Deuterium oxide
Formula: D₂O
Boiling Pt: 101.4 °C (760 mmHg)
Melting Pt: 3.8 °C
Density: 1.105 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00044636
CAS Number: 7789-20-0

Order Now

Specification Test Results

Appearance Clear colorless Liquid
D-Enrichment ≥99.75 %
Water ≥30.0 %
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR