2-(Diphenylphosphino)benzaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (2-Formylphenyl)diphenylphosphine

42640-0010
TS42640-0010EA 78 USD
TS42640-0010
2-(Diphenylphosphino)benzaldehyde 97%
2-(Diphenylphosphino)benzaldehyde
Formula: C₁₉H₁₅OP
Melting Pt: 112…115 °C
Storage Temperature: Ambient
MDL Number: MFCD00013367
CAS Number: 50777-76-9

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 112 to 120 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR