6-Carboxyfluorescein 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3',6'-Dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-6-carboxylic acid, 6-FAM

42580-1000 42580-5000
TS42580-1000EA 136 USD
TS42580-1000 TS42580-5000
6-Carboxyfluorescein 96%
6-Carboxyfluorescein
Formula: C₂₁H₁₂O₇
MW: 376.32 g/mol
Melting Pt: >300 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00036873
CAS Number: 3301-79-9

Order Now

Specification Test Results

Appearance Yellow to orange Powder
Infrared spectrum Conforms
HPLC ≥95 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR