Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.8% ACS

Supplier: Thermo Scientific Chemicals

Synonyms: 2-Amino-2-(hydroxymethyl)propane-1,3-diol(tris), Tromethamine, Tris(hydroxymethyl)methylamine, 2-Amino-2-(hydroxymethyl)propane-1,3-diol, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, Trizma(R) base, TRIS base, THAM

42457-5000 42457-0025 42457-1000
TS42457-5000EA 128 USD
TS42457-5000 TS42457-0025 TS42457-1000
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.8% ACS
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)
Formula: NH₂C(CH₂OH)₃
MW: 121.14 g/mol
Boiling Pt: 357
Melting Pt: >168.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00004679
CAS Number: 77-86-1
EINECS: 201-064-4
Merck Index: 13,09842
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11a03d1ab6b2c11b9a2c83f0a73fc4264cec0110&vid=5ce890cdf298a59a7900a07fee1c857a5e5e61e0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Water ≤2 % (K.F.)
UV at 290 nm A: ≤0.2
Titration with HCl 99.8 to 100.1 % (On dry basis)
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR