Sulfuric acid 95% in water ACS

Supplier: Thermo Scientific Chemicals
Danger

424520025 424520026 424520100 424525000 424525001
TS424520025EA 154 USD
TS424520025 TS424520026 TS424520100 TS424525000 TS424525001
Sulfuric acid 95% in water ACS
Sulfuric acid
Formula: H₂SO₄
MW: 98.08
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.83 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8 II

Order Now

Specification Test Results

Appearance Free from suspended or insoluble matter
Color scale ≤10 APHA
Residue after ignition ≤0.0005 %
Titration with NaOH 95.0 to 98.0 %
Chloride (Cl) ≤0.2 ppm
Nitrate (NO3) ≤0.5 ppm
Ammonium (NH4) ≤2 ppm
Arsenic (As) ≤0.01 ppm
Iron (Fe) ≤0.2 ppm
Mercury (Hg) ≤5 ppb
Heavy metals (as Pb) ≤1 ppm
KMnO4-reducing substance ≤2 ppm (as SO2)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR