Sodium thiosulfate pentahydrate 99.5+% ACS

Supplier: Thermo Scientific Chemicals

AC42446-0010 AC42446-0051
TS42446-0010EA 102 USD
TS42446-0010 TS42446-0051
Sodium thiosulfate pentahydrate 99.5+% ACS
Sodium thiosulphate pentahydrate
Formula: Na₂S₂O₃.5H₂O
Melting Pt: 48.5 °C
Density: 1.72 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149186
CAS Number: 10102-17-7
EINECS: 231-867-5
Merck Index: 12,08844

Order Now

Specification Test Results

Appearance White Crystals
Titration Iodimetric 99.5 to 101.0 %
pH 6.0 to 8.4 (5 % solution at 25.0°C)
Sulfide (S) Passes test (limit about 1 ppm)
SO4 and SO3 (as SO4) ≤0.1 %
Insoluble matter ≤0.005 %
Nitrogen compounds (as N) ≤0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR