Trisodium phosphate dodecahydrate 98+% ACS

Supplier: Thermo Scientific Chemicals
Warning

42440-0025 424405000
TS42440-0025EA 406 USD
TS42440-0025 TS424405000
Trisodium phosphate dodecahydrate 98+% ACS
tri-Sodium phosphate dodecahydrate
Formula: Na₃PO₄·12H₂O
Melting Pt: 75 °C
Density: 1.62 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149198
CAS Number: 10101-89-0
EINECS: 231-509-8
REACH: 01-2119489800-32-xxxx
Merck Index: 13,08736

Order Now

Specification Test Results

Appearance Colorless to white Powder or crystals or needles
Titration excess HCl 98.0 to 102.0 %
Chloride (Cl) ≤10 ppm
Sulfate (SO4) ≤100 ppm
Iron (Fe) ≤10 ppm
Heavy metals (as Pb) ≤10 ppm
Excess alkali (as NaOH) ≤2.5 %
Insoluble matter ≤0.01 %
Methanol ≤0.05 %
Peroxides ≤1 mg/kg H2O2


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR