Sodium hydroxide 97+%, pellets ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

42433-0025 42433-0100 42433-5000 42433-0251 424330010 424330250
TS42433-0025EA 186 USD
TS42433-0025 TS42433-0100 TS42433-5000 TS42433-0251 TS424330010 TS424330250
Sodium hydroxide 97+%, pellets ACS
Sodium hydroxide
Formula: NaOH
Boiling Pt: 1390 °C (760 mmHg)
Melting Pt: 318 °C
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8,II
REACH: 01-2119457892-27
Merck Index: 14,08627

Order Now

Specification Test Results

Appearance White Pellets
Titration with HCl ≥97.0 %
Chloride (Cl) ≤50 ppm
Phosphate (PO4) ≤10 ppm
Sulfate (SO4) ≤30 ppm
Sodium carbonate (Na2CO3) ≤1.0 %
Calcium (Ca) ≤50 ppm
Iron (Fe) ≤10 ppm
Magnesium (Mg) ≤20 ppm
Mercury (Hg) ≤0.1 ppm
Nickel (Ni) ≤10 ppm
Potassium (K) ≤200 ppm
Heavy metals (as Ag) ≤20 ppm
Nitrogen compounds (as N) ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR