Sodium acetate trihydrate 99+% ACS
Supplier: Thermo Scientific Chemicals
Synonyms:
Acetic acid sodium salt trihydrate
AC42426-0010
AC42426-0025
AC42426-0250
TS42426-0010EA
72
USD
TS42426-0010
TS42426-0025
TS42426-0250
Sodium acetate trihydrate 99+% ACS
Sodium acetate trihydrate
Formula:
H₃CCOONa·3H₂O Melting Pt: 58 °C Storage Temperature: Ambient |
MDL Number:
MFCD00071557 CAS Number: 6131-90-4 EINECS: 204-823-8 Merck Index: 13,08642 |
|
Specification Test Results
Appearance | Transparent white Crystals |
Infrared spectrum | Conforms |
pH | 7.5 to 9.2 (5 % at 25.0°C) |
Titration with HClO4 | 99.0 to 101.0 % |
Chloride (Cl) | ≤0.001 % |
Phosphate (PO4) | ≤5 ppm |
Sulfate (SO4) | ≤0.002 % |
Calcium (Ca) | ≤0.005 % |
Iron (Fe) | ≤5 ppm |
Magnesium (Mg) | ≤0.002 % |
Potassium (K) | ≤0.005 % |
Heavy metals (as Pb) | ≤5 ppm |
Insoluble matter | ≤0.005 % |
KMnO4-reducing substance | Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...