Potassium sulfate 99+% for biochemistry
Supplier: Thermo Scientific Chemicals
AC42421-0010
AC42421-2500
TS42421-0010EA
183
USD
TS42421-0010
TS42421-2500
Potassium sulfate 99+% for biochemistry
Potassium sulphate
Formula:
K₂SO₄ MW: 174.26 g/mol Boiling Pt: 1698 Melting Pt: 1069 Storage Temperature: Ambient |
MDL Number:
MFCD00011388 CAS Number: 7778-80-5 EINECS: 231-915-5 Merck Index: 13,07759 |
Specification Test Results
Appearance | White Crystals |
Solubility | (10 g in 150 ml water) Clear colorless |
pH | 5.5 to 8.5 (5 % in water) |
Titration after Ion exchange | ≥99.0 % |
Chloride (Cl) | ≤10 ppm |
Calcium (Ca) | ≤0.01 % |
Iron (Fe) | ≤5 ppm |
Magnesium (Mg) | ≤0.002 % |
Sodium (Na) | ≤0.02 % |
Heavy metals (as Pb) | ≤5 ppm |
Insoluble matter | ≤0.1 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...