Potassium hexacyanoferrate(II) trihydrate 98.5% ACS
Supplier: Thermo Scientific Chemicals
42413-0025
AC42413-0050
AC42413-5000
TS42413-0025EA
295
USD
TS42413-0025
TS42413-0050
TS42413-5000
Potassium hexacyanoferrate(II) trihydrate 98.5% ACS
Potassium hexacyanoferrate (II) trihydrate
Formula:
K₄Fe(CN)₆·3H₂O Melting Pt: 70 °C Density: 1.85 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00167023 CAS Number: 14459-95-1 EINECS: 237-722-2 Merck Index: 12,07793 |
Specification Test Results
Appearance | Yellow Crystalline powder |
Titration with KMnO4 | 98.5 to 102.0 % |
Chloride (Cl) | ≤100 ppm |
Sulfate (SO4) | Passes test (limit about 0.01%) |
Insoluble matter | ≤0.005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...