Potassium carbonate, anhydrous 99+% ACS

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Potash, Carbonic acid potassium salt, Carbonic acid, dipotassium salt

AC42408-0025 AC42408-0100 AC42408-1000 AC42408-5000
TS42408-0025EA 358 USD
TS42408-0025 TS42408-0100 TS42408-1000 TS42408-5000
Potassium carbonate, anhydrous 99+% ACS
Potassium carbonate
Formula: K₂CO₃
MW: 138.21 g/mol
Melting Pt: 891
Storage Temperature: Ambient
MDL Number: MFCD00011382
CAS Number: 584-08-7
EINECS: 209-529-3
Merck Index: 13,07702
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d5900faa1ba52c3213eed8f6bb2d952bac1f30f&vid=6298361525e6af416556b5694c74a8a91faa437c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Granular powder
Titration with HCl ≥99.0 %
Chloride (Cl) ≤0.003 %
Phosphate (PO4) ≤0.001 %
Calcium (Ca) ≤0.005 %
Iron (Fe) ≤5 ppm
Magnesium (Mg) ≤0.002 %
Sodium (Na) ≤0.02 %
Heavy metals (ICP-OES) ≤5 ppm
Silicon dioxide (SiO2) ≤0.005 %
Sulfur compounds (as SO4) ≤0.004 %
Insoluble matter ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR