D-(-)-Mannitol ACS

Supplier: Thermo Scientific Chemicals

42392-0010 42392-0050 42392-2500
TS42392-0010EA 177 USD
TS42392-0010 TS42392-0050 TS42392-2500
D-(-)-Mannitol ACS
D(-)-Mannitol
Formula: HOH₂C(CH(OH))₄CH₂OH
Melting Pt: 165…170 °C
Storage Temperature: Ambient
MDL Number: MFCD00064287
CAS Number: 69-65-8
EINECS: 200-711-8
Merck Index: 13,05769
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=163f556dca45086e05ff490ddc8eeb65d04aea05&vid=408e3f5566e309064ce68988cdd160305c5b8c0a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.05 % (5 g, 105°C, 2 hrs)
Residue after ignition ≤0.01 %
Specific optical rotation +23.3° to +24.3° (25°C, 589 nm)
Heavy metals (ICP-OES) ≤5 ppm (as Pb)
Titrable acid ≤0.0008 meq/g
Insoluble matter ≤0.01 %
Reducing sugars Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR