Lead(II) nitrate ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
42385-5000
42385-0025
42385-0050
TS42385-5000EA
106
USD
TS42385-5000
TS42385-0025
TS42385-0050
Lead(II) nitrate ACS
Lead (II) nitrate
Formula:
Pb(NO₃)₂ Melting Pt: 470 °C (dec.) Density: 4.53 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00011153 CAS Number: 10099-74-8 EINECS: 233-245-9 UN: 1469 ADR: 5.1,II Merck Index: 13,05432 |
Specification Test Results
Appearance | White Crystalline powder |
Titration Complexometric | ≥99.0 % |
Chloride (Cl) | ≤10 ppm |
Calcium (Ca) | ≤50 ppm |
Copper (Cu) | ≤20 ppm |
Iron (Fe) | ≤10 ppm |
Potassium (K) | ≤50 ppm |
Sodium (Na) | ≤200 ppm |
Insoluble matter | ≤0.005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...