Hydrazinium sulfate ACS

Supplier: Thermo Scientific Chemicals
Danger

42377-0500 42377-1000 42377-5000
TS42377-0500EA 60 USD
TS42377-0500 TS42377-1000 TS42377-5000
Hydrazinium sulfate ACS
Hydrazinium sulphate
Formula: NH₂NH₂·H₂SO₄
Melting Pt: 254 °C
Storage Temperature: Ambient
MDL Number: MFCD00044873
CAS Number: 10034-93-2
EINECS: 233-110-4
UN: 3288
ADR: 6.1,III
Merck Index: 14,04772
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e277940036b3cf71839ff7458f0c7d08bff6f6d5&vid=7a677e6f1725cfa1b9bdef05cc04304e6739079e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline powder
Titration Iodimetric ≥99.0 %
Residue after ignition ≤0.05 %
Chloride (Cl) ≤50 ppm
Iron (Fe) ≤10 ppm
Heavy metals (as Pb) ≤20 ppm
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR