Formamide 99.6% ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Carbamaldehyde

42374-5000 423740010 423741000
TS42374-5000EA 197 USD
TS42374-5000 TS423740010 TS423741000
Formamide 99.6% ACS
Formamide
Formula: HCONH₂
Boiling Pt: 210 °C (1013 hPa)
Melting Pt: 2…3 °C
Density: 1.133 g/cm³
Flash Pt: 175 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007941
CAS Number: 75-12-7
EINECS: 200-842-0
Merck Index: 13,04261
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f381691a96baf5054ca8291e1fa9e9603b687e31&vid=e54dda1a985d7a4f9485c2cc5233498087e43f88&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
GC ≥99.5 %
Color scale ≤10 APHA
Freezing point 2.0 to 3.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR