Ammonium iron (II) sulfate hexahydrate 99% ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
di-Ammonium iron (II) sulfate hexahydrate, Ferrous Ammonium sulphate Hexahydrate, Ferrous ammonium sulfate hexahydrate, Ferrous Ammonium sulfate Hexahydrate, di-Ammonium iron (II) sulphate hexahydrate, Ferrous ammonium sulphate hexahydrate, Ammonium iron(Ⅱ) sulphate hexahydrate, Ammonium iron(Ⅱ) sulfate hexahydrate, Ammonium ferrous sulphate hexahydrate, Ammonium iron(II) sulphate hydrate, Ammonium ferrous sulfate hexahydrate, Ammonium iron(II) sulfate hydrate
AC42372-0030
AC42372-1000
AC42372-5000
TS42372-0030EA
598
USD
TS42372-0030
TS42372-1000
TS42372-5000
Ammonium iron (II) sulfate hexahydrate 99% ACS
Ammonium iron (II) sulphate hexahydrate
Formula:
(NH₄)₂Fe(SO₄)₂·6H₂O Melting Pt: 100 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00150530 CAS Number: 7783-85-9 EINECS: 233-151-8 |
Specification Test Results
Appearance | Blue-green Crystals |
Titration with KMnO4 | 98.5 to 101.5 % |
Phosphate (PO4) | ≤0.003 % |
Calcium (Ca) | ≤0.005 % |
Copper (Cu) | ≤0.003 % |
Magnesium (Mg) | ≤0.002 % |
Manganese (Mn) | ≤0.01 % |
Potassium (K) | ≤0.002 % |
Sodium (Na) | ≤0.02 % |
Zinc (Zn) | ≤0.003 % |
Ferric iron (Fe 3+) | ≤100 ppm (May increase due to inherent oxidation on storage) |
Insoluble matter | ≤0.01 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...