Iron(III) nitrate nonahydrate 98+% ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ferric nitrate nonahydrate, Iron trinitrate nonahydrate, Iron(III) nitrate hydrate, Ferric Nitrate, 9-Hydrate
42371-0030
42371-5000
AC42371-0100
TS42371-0030EA
282
USD
TS42371-0030
TS42371-5000
TS42371-0100
Iron(III) nitrate nonahydrate 98+% ACS
Iron (III) nitrate nonahydrate
Formula:
Fe(NO₃)₃·9H₂O MW: 404 g/mol Boiling Pt: 125 Melting Pt: 47 Density: 1.68 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00149708 CAS Number: 7782-61-8 EINECS: 233-899-5 UN: 1466 ADR: 5.1 III Merck Index: 13,04057 |
Specification Test Results
Appearance | Almost white to light gray to light purple Adhering crystals or chunks |
Titration Complexometric | 98.0 to 101.0 % |
Chloride (Cl) | ≤5 ppm |
Sulfate (SO4) | ≤100 ppm |
Calcium (Ca) | ≤100 ppm |
Magnesium (Mg) | ≤50 ppm |
Potassium (K) | ≤50 ppm |
Sodium (Na) | ≤500 ppm |
Insoluble matter | ≤0.005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...