Iron(III) chloride hexahydrate 97+% ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Iron trichloride hexahydrate, Iron trichloride hexahydrate, Iron (III) chloride 6-hydrate, Iron trichloride 6-hydrate, Ferric chloride 6-hydrate, Ferric chloride hexahydrate, Ferric chloride hexahydrate

42370-5000 AC42370-0025 AC42370-0050 AC42370-0100
TS42370-5000EA 125 USD
TS42370-5000 TS42370-0025 TS42370-0050 TS42370-0100
Iron(III) chloride hexahydrate 97+% ACS
Iron (III) chloride hexahydrate
Formula: FeCl₃·6H₂O
Boiling Pt: 280…285 °C
Melting Pt: 37 °C
Storage Temperature: Ambient
MDL Number: MFCD00149712
CAS Number: 10025-77-1
EINECS: 231-729-4
UN: 3260
ADR: 8,III
Merck Index: 13,04048
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=808c75376e55f29296d58eb1e5a1e184039ce733&vid=6d820873011bee3ff401441544f7f85599e40626&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Yellow to orange to light brown Adhering powder and/or lumps
Titration Iodometric 97.0 to 102.0 %
Nitrate (NO3) ≤100 ppm
Sulfate (SO4) ≤100 ppm
Calcium (Ca) ≤100 ppm
Copper (Cu) ≤30 ppm
Magnesium (Mg) ≤50 ppm
Potassium (K) ≤50 ppm
Sodium (Na) ≤500 ppm
Zinc (Zn) ≤30 ppm
Ferrous iron (Fe 2+) Passes test (limit about 0.002%)
Phosphorous compounds (as PO4) ≤0.01 %
Insoluble matter ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR