Diphenylamine ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
N-Phenylaniline, N-Phenylbenzeneamine
42365-0050
42365-1000
42365-5000
TS42365-0050EA
54
USD
TS42365-0050
TS42365-1000
TS42365-5000
Diphenylamine ACS
Diphenylamine
Formula:
(C₆H₅)₂NH MW: 169.23 g/mol Boiling Pt: 302 Melting Pt: 52…54 °C Flash Pt: 152 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00003014 CAS Number: 122-39-4 EINECS: 204-539-4 UN: 3077 ADR: 9 III Merck Index: 13,03349 |
Specification Test Results
Appearance | White to off-white Crystalline powder or crystals or flakes |
Infrared spectrum | Conforms |
Melting point | 52.5 to 54.0 °C |
Solubility | (in alcohol) Passes test |
Residue after ignition | ≤0.03 % |
Sensitivity | (to nitrate) Passes test |
Nitrate (NO3) | Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...