Ammonium cerium(IV) nitrate 99% ACS

Supplier: Thermo Scientific Chemicals
Danger

42354-0025 42354-1000 42354-5000 AC42354-0250 AC42354-0500
TS42354-0025EA 920 USD
TS42354-0025 TS42354-1000 TS42354-5000 TS42354-0250 TS42354-0500
Ammonium cerium(IV) nitrate 99% ACS
Ammonium cerium (IV) nitrate
Formula: (NH₄)₂Ce(NO₃)₆
Melting Pt: 107…108 °C
Storage Temperature: Ambient
MDL Number: MFCD00151121
CAS Number: 16774-21-3
EINECS: 240-827-6
UN: 3085
ADR: 5.1,II
Merck Index: 13,00509

Order Now

Specification Test Results

Appearance Orange Crystalline powder or crystals
Titration ≥98.5 %
Chloride (Cl) ≤0.01 %
Phosphate (PO4) ≤0.02 %
Iron (Fe) ≤0.005 %
Insoluble matter ≤0.05 % (in dilute sulfuric acid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR