Orthoboric acid 99.6% ACS

Supplier: Thermo Scientific Chemicals
Danger

42348-5000 423480020
TS42348-5000EA 96 USD
TS42348-5000 TS423480020
Orthoboric acid 99.6% ACS
ortho-Boric acid
Formula: B(OH)₃
Boiling Pt: 185 °C (1013 hPa)
Melting Pt: 169 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011337
CAS Number: 10043-35-3
EINECS: 233-139-2
REACH: 01-2119486683-25
Merck Index: 13,01326
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=098dd700154daaf6338ebb270902bb2bc704f27d&vid=6eec7d529090dd4c4b966ed8cd8d057153c5a47b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline powder
Loss on drying ≤0.5 %
Titration with NaOH ≥99.5 %
Chloride (Cl) ≤0.001 %
Phosphate (PO4) ≤0.001 %
Sulfate (SO4) ≤0.01 %
Calcium (Ca) ≤0.005 %
Iron (Fe) ≤0.001 %
Heavy metals (as Pb) ≤0.001 %
Insoluble matter ≤0.005 % (in methanol)
Non-volatiles with MeOH ≤0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR