Barium acetate 99+% ACS

Supplier: Thermo Scientific Chemicals
Warning

AC42343-0025 AC42343-1000 AC42343-5000
TS42343-0025EA 330 USD
TS42343-0025 TS42343-1000 TS42343-5000
Barium acetate 99+% ACS
Barium acetate
Formula: (H₃CCOO)₂Ba
Melting Pt: ∼450 °C
Density: 2.46 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00012447
CAS Number: 543-80-6
EINECS: 208-849-0
UN: 1564
ADR: 6.1,III
Merck Index: 13,00968

Order Now

Specification Test Results

Appearance White Crystalline powder
Titration Complexometric 99.0 to 102.0 %
Chloride (Cl) ≤0.001 %
Calcium (Ca) ≤0.05 %
Iron (Fe) ≤0.001 %
Potassium (K) ≤0.03 %
Sodium (Na) ≤0.05 %
Strontium (Sr) ≤0.2 %
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.01 %
Oxidizing substance (as NO3) ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR