Ammonium nitrate 98% ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
42335-0010
AC42335-0250
TS42335-0010EA
135
USD
TS42335-0010
TS42335-0250
Ammonium nitrate 98% ACS
Ammonium nitrate
Formula:
NH₄NO₃ Boiling Pt: 230 °C (1013 hPa) Melting Pt: 169 °C Density: 1.72 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00011425 CAS Number: 6484-52-2 EINECS: 229-347-8 UN: 1942 ADR: 5.1,III Merck Index: 13,00538 |
Specification Test Results
Appearance | White Adhering crystals |
Residue after ignition | ≤0.01 % |
pH | 4.5 to 6.0 (5 % at 25°C) |
Titration excess NaOH | ≥97.5 % |
Chloride (Cl) | ≤5 ppm |
Nitrite (NO2) | (Limit about 5 ppm) Passes test |
Phosphate (PO4) | ≤5 ppm |
Sulfate (SO4) | ≤0.002 % |
Iron (Fe) | ≤2 ppm |
Heavy metals (as Pb) | ≤5 ppm |
Insoluble matter | ≤0.005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...