Acetonitrile 99.6% ACS

Supplier: Thermo Scientific Chemicals
Danger

423250010 423250025 423250100 423255000
TS423250010EA 135 USD
TS423250010 TS423250025 TS423250100 TS423255000
Acetonitrile 99.6% ACS
Acetonitrile
Formula: H₃CCN
Boiling Pt: 81…82 °C (760 mmHg)
Melting Pt: –46 °C
Density: 0.781 g/cm³
Flash Pt: 2 °C
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
REACH: 01-2119471307-38

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.3 % (K.F.)
GC ≥99.5 %
Color scale ≤10 APHA
Residue after evaporation ≤0.005 %
Titrable acid ≤0.008 meq/g
Titrable base ≤0.0006 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR