1-Pentanesulfonic acid sodium salt monohydrate 98+% for HPLC
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Sodium 1-pentanesulfonate monohydrate, Sodium 1-pentanesulphonate monohydrate
AC42320-0250
AC42320-1000
TS42320-0250EA
295
USD
TS42320-0250
TS42320-1000
1-Pentanesulfonic acid sodium salt monohydrate 98+% for HPLC
1-Pentanesulphonic acid sodium salt monohydrate
Formula:
C₅H₁₁NaO₃S·1H₂O MW: 192.21 Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007541 CAS Number: 207605-40-1 EINECS: 245-208-4 |
Specification Test Results
Appearance | Colorless to white Powder or crystals or flakes |
Infrared spectrum | Conforms |
UV | at 260 nm A: ≤0.01 |
Titration after Ion exchange | ≥98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...