2-Chloroethylammonium chloride 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Chloroethylamine hydrochloride, 2-Chloroethylamine HCl, 2-Chloroethanamine hydrochloride
42286-0010
42286-0250
42286-1000
TS42286-0010EA
347
USD
TS42286-0010
TS42286-0250
TS42286-1000
2-Chloroethylammonium chloride 98%
2-Chloroethylammonium chloride
Formula:
C₂H₇Cl₂N MW: 115.99 g/mol Melting Pt: 140…150 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012887 CAS Number: 870-24-6 EINECS: 212-793-2 |
Specification Test Results
Appearance | White to light beige Crystalline powder, crystals or chunks |
Infrared spectrum | Conforms |
Melting point | 140 to 150 °C |
Loss on drying | ≤1 % |
Titration with HClO4 | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...