p-Xylene 99% for spectroscopy

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,4-Dimethylbenzene

42267-0010
TS42267-0010EA 356 USD
TS42267-0010
p-Xylene 99% for spectroscopy
p-Xylene
Formula: C₆H₄(CH₃)₂
MW: 106.17 g/mol
Boiling Pt: 138.3 °C (1013 hPa)
Melting Pt: 13 °C
Density: 0.866 g/cm³
Flash Pt: 25 °C
Storage Temperature: Ambient
MDL Number: MFCD00008556
CAS Number: 106-42-3
EINECS: 203-396-5
UN: 1307
ADR: 3,III
Merck Index: 12,10214
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d8b04563a5941cefd103ebd3048bcf8486f2bdac&vid=e5db83ad1f0cb6a583610c58cea291dcbe1be1f3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
GC ≥98.5 %
UV at 420 nm A: ≤0.01
Color scale ≤15 APHA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR