Tris(2-methylphenyl)phosphine 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tris(o-tolyl)phosphine, Tri-o-tolylphosphine

42232-0010 42232-0050 42232-0250
TS42232-0010EA 29 USD
TS42232-0010 TS42232-0050 TS42232-0250
Tris(2-methylphenyl)phosphine 99%
Tris(2-methylphenyl)phosphine
Formula: C₂₁H₂₁P
Melting Pt: 126…127 °C
Storage Temperature: Ambient
MDL Number: MFCD00008514
CAS Number: 6163-58-2
EINECS: 228-193-9

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 122 to 126 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR