Tricyclohexylphosphine 97%

Supplier: Thermo Scientific Chemicals
Warning

42161-0050 42161-0010 42161-0250
TS42161-0050EA 93 USD
TS42161-0050 TS42161-0010 TS42161-0250
Tricyclohexylphosphine 97%
Tricyclohexylphosphine
Formula: C₁₈H₃₃P
Melting Pt: 77…83 °C
Storage Temperature: Ambient
MDL Number: MFCD00003853
CAS Number: 2622-14-2
EINECS: 220-069-2

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 77 to 83 °C
NMR (31P-NMR) ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR