D-α-Tocopherol 97+%

Supplier: Thermo Scientific Chemicals

Synonyms: (+)-α-Tocopherol, Vitamin E Alcohol, (R)-2,5,7,8-Tetramethyl-2-((4R,8R)-4,8,12-trimethyltridecyl)chroman-6-ol, α-Vitamin E, 3,4-Dihydro-2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-2H-benzopyran-6-ol, Vitamin E, D(+)-α-Tocopherol

42103-1000
TS42103-1000EA 179 USD
TS42103-1000
D-α-Tocopherol 97+%
D-α-Tocopherol
Formula: C₂₉H₅₀O₂
MW: 430.71 g/mol
Boiling Pt: 200…220 °C (.1 mmHg)
Melting Pt: 2.5…3.5 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00072045
CAS Number: 59-02-9
EINECS: 233-466-0

Order Now

Specification Test Results

Appearance Clear yellow to red-brown Viscous liquid
Infrared spectrum Conforms
Refractive index 1.5050 to 1.5070 (20°C, 589 nm)
GC ≥97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR