Thallium(III) trifluoroacetate tech.

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Thallic trifluoroacetate

AC42076-0250 AC42076-1000
TS42076-0250EA 236 USD
TS42076-0250 TS42076-1000
Thallium(III) trifluoroacetate tech.
Thallium (III) trifluoroacetate
Formula: C₆F₉O₆Tl
MW: 543.43 g/mol
Boiling Pt: 160
Melting Pt: 120.5
Storage Temperature: Ambient
MDL Number: MFCD00000414
CAS Number: 23586-53-0
EINECS: 245-761-1
UN: 1707
ADR: 6.1 II

Order Now

Specification Test Results

Appearance Light yellow to beige Powder
Infrared spectrum Conforms
Melting point 56 to 60 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR