N,N,N',N'-Tetramethylethylenediamine (TEMED) 99% for biochemistry, for electrophoresis

Supplier: Thermo Scientific Chemicals
Danger

42058-0050 42058-0025 42058-0500 42058-1000 42058-5000
TS42058-0050EA 24 USD
TS42058-0050 TS42058-0025 TS42058-0500 TS42058-1000 TS42058-5000
N,N,N',N'-Tetramethylethylenediamine (TEMED) 99% for biochemistry, for electrophoresis
N,N,N',N'-Tetramethylethylenediamine (TEMED)
Formula: C₆H₁₆N₂
Boiling Pt: 120 °C (1013 hPa)
Melting Pt: –55 °C
Density: 0.77 g/cm³
Flash Pt: 17 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008335
CAS Number: 110-18-9
EINECS: 203-744-6
UN: 2372
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4169 to 1.4189 (20 °C, 589 nm)
Solubility (10 % in water) Clear and no haze
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR