Tetramethylammonium hydroxide 25% in water

Supplier: Thermo Scientific Chemicals
Danger

42052-0050 420520010 420521000
TS42052-0050EA 741 USD
TS42052-0050 TS420520010 TS420521000
Tetramethylammonium hydroxide 25% in water
Tetramethylammonium hydroxide
Formula: C₄H₁₃NO
MW: 91.15
Boiling Pt: 110 °C (1013 hPa)
Melting Pt: –25 °C
Density: 1.014 g/cm³
Flash Pt: >95 °C
Storage Temperature: Ambient
MDL Number: MFCD00008280
CAS Number: 75-59-2
EINECS: 200-882-9
UN: 1835
ADR: 8 II
Merck Index: 14,09224

Order Now

Specification Test Results

Appearance Clear colorless to yellow Solution
Refractive index 1.3780 to 1.3840 (20°C, 589 nm)
Titration with HCl 23 to 27 %
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR