Sodium iodate 99%, powder, Extra Pure reagent

Supplier: Thermo Scientific Chemicals
Danger

AC41956-0050 AC41956-5000
TS41956-0050EA 27 USD
TS41956-0050 TS41956-5000
Sodium iodate 99%, powder, Extra Pure reagent
Sodium iodate
Formula: INaO₃
Melting Pt: 560 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00003533
CAS Number: 7681-55-2
EINECS: 231-672-5
UN: 1479
ADR: 5.1,II
Merck Index: 13,08704
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=806173453dcec1d2f791367d269d662d3459149a&vid=16d5efbf45a7cc1cd6399272211e2455e47a2e4b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Titration Iodometric ≥98.5 %
pH 5.0 to 8.0 (5 % at 20°C)
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR