Sodium dodecyl sulfate (SDS) 98% for biochemistry, for electrophoresis
Supplier: Thermo Scientific Chemicals
|
Danger
|
41953-0010
41953-0050
41953-0250
41953-1000
TS41953-0010EA
432
USD
TS41953-0010
TS41953-0050
TS41953-0250
TS41953-1000
Sodium dodecyl sulfate (SDS) 98% for biochemistry, for electrophoresis
Sodium dodecyl sulphate (SDS)
Formula:
CH₃(CH₂)₁₁OSO₃Na Melting Pt: 206 °C Flash Pt: >150 °C Storage Temperature: Ambient |
MDL Number:
MFCD00036175 CAS Number: 151-21-3 EINECS: 205-788-1 UN: 1325 ADR: 4.1,III Merck Index: 14,08636 |
|
Specification Test Results
Appearance | White Crystalline powder or flakes |
Infrared spectrum | Conforms |
Loss on drying | ≤1 % (1 g, 105°C, 3 h) |
Solubility | (3 % in water) Clear colorless |
GC | ≥97.5 % |
UV | at 280 nm A: ≤0.04 |
Chloride (Cl) | ≤500 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...