Sodium borohydride 98%, granules ∼ 3 mm
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Sodium tetrahydridoborate, Sodium tetrahydroborate
41947-1000
AC41947-5000
TS41947-1000EA
86
USD
TS41947-1000
TS41947-5000
Sodium borohydride 98%, granules ∼ 3 mm
Sodium borohydride
Formula:
NaBH₄ MW: 37.83 g/mol Melting Pt: 400 (dec.) Flash Pt: 69 Storage Temperature: Ambient |
MDL Number:
MFCD00003518 CAS Number: 16940-66-2 EINECS: 241-004-4 UN: 1426 ADR: 4.3 I Merck Index: 13,08664 |
Specification Test Results
Appearance | White Granules |
Titration Iodometric | ≥97.5 % |
Water | ≤0.2 % (Coulometric) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...