Sodium hydrogen sulfate monohydrate 97%, crystals, Extra Pure reagent

Supplier: Thermo Scientific Chemicals
Danger

AC41943-0025 AC41943-0100 AC41943-5000
TS41943-0025EA 351 USD
TS41943-0025 TS41943-0100 TS41943-5000
Sodium hydrogen sulfate monohydrate 97%, crystals, Extra Pure reagent
Sodium hydrogen sulphate monohydrate
Formula: NaHSO₄·H₂O
Melting Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00149210
CAS Number: 10034-88-5
EINECS: 231-665-7
Merck Index: 13,08658

Order Now

Specification Test Results

Appearance White Adhering crystals
Titration with NaOH ≥96.0 %
Chloride (Cl) ≤20 ppm
Iron (Fe) ≤50 ppm
Heavy metals (as Pb) ≤50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR