Resazurin sodium salt, pure, certified

Supplier: Thermo Scientific Chemicals
Warning

41890-0050 41890-0250 41890-0010
TS41890-0050EA 84 USD
TS41890-0050 TS41890-0250 TS41890-0010
Resazurin sodium salt, pure, certified
Resazurin sodium salt
Formula: C₁₂H₆NNaO₄
Storage Temperature: Ambient
MDL Number: MFCD00005036
CAS Number: 62758-13-8

Order Now

Specification Test Results

Appearance Dark green to black Powder
Infrared spectrum Conforms
UV spectrum Complies
Dye content ≥80 %
BSC Certification Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR