Phenolphthalein ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one
41718-0026
41718-1000
41718-5000
TS41718-0026EA
422
USD
TS41718-0026
TS41718-1000
TS41718-5000
Phenolphthalein ACS
Phenolphthalein
Formula:
C₂₀H₁₄O₄ Melting Pt: 258…263 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005913 CAS Number: 77-09-8 EINECS: 201-004-7 REACH: 01-2119498295-24 Merck Index: 14,07243 |
|
Specification Test Results
Appearance | Off-white Crystalline powder |
Infrared spectrum | Conforms |
Visual transition interval | From pH 8.0 (colorless) to pH 10 (red) |
Clarity of alcohol solution | Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...