Phenolphthalein ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one

41718-0026 41718-1000 41718-5000
TS41718-0026EA 422 USD
TS41718-0026 TS41718-1000 TS41718-5000
Phenolphthalein ACS
Phenolphthalein
Formula: C₂₀H₁₄O₄
Melting Pt: 258…263 °C
Storage Temperature: Ambient
MDL Number: MFCD00005913
CAS Number: 77-09-8
EINECS: 201-004-7
REACH: 01-2119498295-24
Merck Index: 14,07243
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d81b539a32ba7c40a473e6afbb8e0dae525d637&vid=91bf141a7ea82baf5df563f73967884d2a4b5d79&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Off-white Crystalline powder
Infrared spectrum Conforms
Visual transition interval From pH 8.0 (colorless) to pH 10 (red)
Clarity of alcohol solution Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR