Nickel (II) oxide 97%, powder

Supplier: Thermo Scientific Chemicals
Danger

AC41558-1000 AC41558-5000
TS41558-1000EA 75 USD
TS41558-1000 TS41558-5000
Nickel (II) oxide 97%, powder
Nickel (II) oxide
Formula: NiO
Melting Pt: 1960 °C
Storage Temperature: Ambient
MDL Number: MFCD00011145
CAS Number: 1313-99-1
EINECS: 215-215-7

Order Now

Specification Test Results

Appearance Green Powder
Titration Complexometric ≥96.0 %
Calcium (Ca) ≤0.003 %
Cobalt (Co) ≤0.025 %
Copper (Cu) ≤0.0005 %
Iron (Fe) ≤0.005 %
Magnesium (Mg) ≤0.01 %
Manganese (Mn) ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR