Methyl (R)-(+)-lactate 98%

Supplier: Thermo Scientific Chemicals
Warning

41449-0050 41449-0250
TS41449-0050EA 75 USD
TS41449-0050 TS41449-0250
Methyl (R)-(+)-lactate 98%
Methyl (R)-(+)-lactate
Formula: C₄H₈O₃
Boiling Pt: 144 °C (1013 hPa)
Density: 1.09 g/cm³
Flash Pt: 49 °C
Storage Temperature: Ambient
MDL Number: MFCD00004517
CAS Number: 17392-83-5
EINECS: 241-420-6
UN: 3272
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4110 to 1.4150 (20°C, 589 nm)
GC ≥97.5 %
Optical rotation +6.5° to +10.5° (20°C, 589 nm) (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR