DL-Lysine monohydrochloride 99%

Supplier: Thermo Scientific Chemicals

Synonyms: (±)-2,6-Diaminocaproic acid monohydrochloride, H-DL-Lys-OH · HCl, H-DL-Lys-OH.HCl, (±)-2,6-Diaminohexanoic acid monohydrochloride, (±)-Lysine monohydrochloride

41337-0010 41337-0250 41337-1000
TS41337-0010EA 864 USD
TS41337-0010 TS41337-0250 TS41337-1000
DL-Lysine monohydrochloride 99%
DL-Lysine monohydrochloride
Formula: C₆H₁₅ClN₂O₂
Melting Pt: 267 °C
Storage Temperature: Ambient
MDL Number: MFCD00064563
CAS Number: 70-53-1
EINECS: 200-739-0

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Specific optical rotation -1° to +1° (20°C, 589 nm) (c=1, water)
Titration with HClO4 98.5 to 101.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR