Lanthanum(III) oxide 91%

Supplier: Thermo Scientific Chemicals

AC41302-1000 AC41302-2500
TS41302-1000EA 88 USD
TS41302-1000 TS41302-2500
Lanthanum(III) oxide 91%
Lanthanum (III) oxide
Formula: La₂O₃
Boiling Pt: 4200 °C (1013 hPa)
Melting Pt: 2315 °C
Storage Temperature: Ambient
MDL Number: MFCD00011071
CAS Number: 1312-81-8
EINECS: 215-200-5

Order Now

Specification Test Results

Appearance White to light gray Powder
Assay ≥89 %
Melting point 170 to 175 °C
GC ≥96.0 %
Titration with NaOH ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR