Isobutanol for HPLC

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: iso-Butyl alcohol, Isobutylalcohol, iso-Butanol, 2-Methyl-1-propanol

412640010
TS412640010EA 170 USD
TS412640010
Isobutanol for HPLC
Isobutanol
Formula: (CH₃)₂CHCH₂OH
MW: 74.12 g/mol
Boiling Pt: 108 °C (760 mmHg)
Melting Pt: –108 °C
Density: 0.8 g/cm³
Flash Pt: 28 °C
Storage Temperature: Ambient
MDL Number: MFCD00004740
CAS Number: 78-83-1
UN: 1212
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.05 %
GC ≥99.45 %
UV at 400 nm A: ≤0.01
Color scale ≤10 APHA
Residue after evaporation ≤0.0005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR