Cetrimonium chloride 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cetyltrimethylammonium chloride, (1-Hexadecyl)trimethylammonium chloride, Hexadecyltrimethylammonium chloride, N-Cetyl-N,N,N-trimethylammonium chloride, N,N,N-Trimethylhexadecan-1-aminium chloride, CTAC

41141-0050 41141-0250 41141-1000 41141-5000
TS41141-0050EA 119 USD
TS41141-0050 TS41141-0250 TS41141-1000 TS41141-5000
Cetrimonium chloride 99%
Cetrimonium chloride
Formula: C₁₉H₄₂ClN
Melting Pt: 234 °C
Storage Temperature: Ambient
MDL Number: MFCD00011773
CAS Number: 112-02-7
EINECS: 203-928-6
UN: 2923
ADR: 8,III

Order Now

Specification Test Results

Appearance White to cream Powder
Infrared spectrum Conforms
Titration Argentometric ≥98.5 %
GC ≥94 %
Specific optical rotation -33.0° to -37.0° (20°C, 589 nm) (c=9 in CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR